missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Owl™ Blotting Paper
Absorb adequate amounts of buffer for semi-dry electroblotting without the loss of mechanical stability with this high-quality blotting paper.
Brand: Thermo Scientific™ FP-2
132.05 GBP valid until 2024-12-31
Use promo code "21973" to get your promotional price.
This item is not returnable.
View return policy
Description
Absorb adequate amounts of buffer for semi-dry electroblotting without the loss of mechanical stability with Thermo Scientific™ Owl™ Blotting Paper, high-quality blotting paper ideal for wicking and blotting applications.
- High purity, 100% cotton fiber
- Available in pre-cut mini gel and large format gel sizes
- Compatible with Owl electroblotting systems
Ordering Information:
- Sold in packs of 100 sheets
Specifications
35 cm | |
Sheets | |
Owl electroblotting systems | |
Sheet | |
35 cm x 45 cm | |
Owl |
45 cm | |
35 mL/min. | |
183 g/m² | |
100% Cotton Fiber | |
0.83 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction