Learn More
Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System Combs
Eliminate comb assembly with Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System Combs.
Brand: Thermo Scientific™ D1-30C
Description
Eliminate comb assembly with Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System Combs. These heavy-duty, one-piece combs are for use with Owl™ D2 Wide-Gel Electrophoresis Systems.
Heavy-duty, one-piece combs eliminate comb assembly.
Specifications
Micro well; 30 (2X) teeth; 1.0mm thick | |
Owl | |
30 (2X) |
Owl™ D2 Wide Gel Electrophoresis System | |
Comb | |
1 mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.