missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Owl™ EasyCast™ B1 Mini Gel Electrophoresis Systems
Cast and run a gel in the same chamber with no tape or additional parts required with the leakproof casting of mini gel electrophoresis systems.
Brand: Thermo Scientific™ B1
This item is not returnable.
View return policy
Description
Cast and run a gel in the same chamber with no tape or additional parts required with the leakproof casting of Thermo Scientific™ Owl™ EasyCast™ B1 Mini Gel Electrophoresis Systems. Durability and ease of use makes this the perfect choice for students and high volume researchers.
Features:
- System offers a wide range of combs.
- Two comb slots on gel tray double sample capacity by doubling number of sample wells.
- UVT gel trays are silk screened with a florescent ruler for easy measurement of bands.
Includes:
- Main buffer chamber
- Thermo Scientific™ SuperSafe™ lid with attached power supply leads
- Thermo Scientific™ EasyCast™ Gasketed UVT gel tray
- Two Combs: 10 & 14 Well, Double-sided, 1.0/1.5mm thick
Ordering Information:
Available with optional buffer exchange ports, (B1-BP), for use with an external recirculation pump.
Warranty and Service Offering:
- Three years
Specifications
CE marked | |
Mini | |
Mini Gel Electrophoresis System | |
600 mL | |
8.63 in. | |
11 cm | |
15 cm | |
3.75 in. | |
150 V |
EasyCast | |
Owl | |
5 to 34 | |
8.63 x 5.88 x 3.75 in. (22 x 15 x 9.5 cm) | |
22 cm | |
5.88 in. | |
9 cm | |
9.5 cm |