Learn More
Thermo Scientific™ Owl™ JGC Gel Casting Systems
Cast polyacrylamide gels faster with Thermo Scientific™ Owl™ JGC Gel Casting Systems, vertical protein gel casters that offer a unique casting method.
Brand: Thermo Scientific™ JGC-2
Code : 04
Additional Details : Weight : 0.80000kg
Description
Cast polyacrylamide gels faster with Thermo Scientific™ Owl™ JGC Gel Casting Systems, vertical protein gel casters that offer a unique casting method.
- Pouch collects excess polyacrylamide and seals bottom of gel, eliminating need for tape or bottom spacers.
- Design allows storage of gels without dehydration by adding buffer and heat sealing.
- System casts from one to four gels.
- Few moving parts result in outstanding durability, extra long life.
Includes:
- Casting stand
- Blocking plate
- Three-point leveling base
- Spacer placer
- 25 gel pouches
Specifications
Gel Electrophoresis | |
20 x 20 cm | |
Gel Caster | |
7.06 in. | |
16 cm | |
17 cm |
Owl JGC Gel Casting System | |
Owl | |
14 cm | |
18 cm | |
6.69 in. |