missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ JGC Gel Casting Systems

Cast polyacrylamide gels faster with Thermo Scientific™ Owl™ JGC Gel Casting Systems, vertical protein gel casters that offer a unique casting method.

Brand:  Thermo Scientific™ JGC-3

Code : 04

Additional Details : Weight : 2.38400kg

 View more versions of this product

Product Code. 11942975

  • £161.00 / Each
Estimated Shipment: 23-09-2024
to see stock.
Add to basket

Description

Description

Cast polyacrylamide gels faster with Thermo Scientific™ Owl™ JGC Gel Casting Systems, vertical protein gel casters that offer a unique casting method.

  • Pouch collects excess polyacrylamide and seals bottom of gel, eliminating need for tape or bottom spacers.
  • Design allows storage of gels without dehydration by adding buffer and heat sealing.
  • System casts from one to four gels.
  • Few moving parts result in outstanding durability, extra long life.

Includes:

  • Casting stand
  • Blocking plate
  • Three-point leveling base
  • Spacer placer
  • 25 gel pouches
Specifications

Specifications

Gel Electrophoresis
20 x 20 cm
Gel Caster
9 in.
20 cm
17 cm
Owl JGC Gel Casting System
Owl
20 cm
23 cm
6.69 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers