missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ JGC Gel Casting Systems

Cast polyacrylamide gels faster with Thermo Scientific™ Owl™ JGC Gel Casting Systems, vertical protein gel casters that offer a unique casting method.

Brand:  Thermo Scientific™ JGC-4

Code : 04

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11952975

  • £124.00 / Each
Estimated Shipment: 24-09-2024
to see stock.
Add to basket

Description

Description

Cast polyacrylamide gels faster with Thermo Scientific™ Owl™ JGC Gel Casting Systems, vertical protein gel casters that offer a unique casting method.

  • Pouch collects excess polyacrylamide and seals bottom of gel, eliminating need for tape or bottom spacers.
  • Design allows storage of gels without dehydration by adding buffer and heat sealing.
  • System casts from one to four gels.
  • Few moving parts result in outstanding durability, extra long life.

Includes:

  • Casting stand
  • Blocking plate
  • Three-point leveling base
  • Spacer placer
  • 25 gel pouches
Specifications

Specifications

Gel Electrophoresis
16 x 14 cm
Gel Caster
5.13 in.
10 cm
13 cm
Owl JGC Gel Casting System
Owl
10 cm
13 cm
5.13 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers