Learn More
Tocris Bioscience™ OXA 06 dihydrochloride
Potent ROCK inhibitor
Brand: Tocris Bioscience™ 5182/10
Additional Details : Weight : 0.00970kg
Description
OXA 06 dihydrochloride is a potent ROCK inhibitor (IC50 = 10 nM). Suppresses pMYPT1 and pCofilin levels in non-small cell lung carcinoma (NSCLC) cell lines. Also inhibits anchorage-independent growth of NSCLC cell lines in vitro.Specifications
>98% | |
10 mg | |
404.31 |
1825455-91-1 | |
OXA 06 dihydrochloride | |
OXA 06 dihydrochloride |
For Research Use Only