missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ OXA 06 dihydrochloride
Potent ROCK inhibitor
£147.94 - £504.31
Specifications
Purity | >98% |
---|---|
CAS | 1825455-91-1 |
Product Type | OXA 06 dihydrochloride |
Molecular Weight (g/mol) | 404.31 |
Inhibitors | OXA 06 dihydrochloride |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18721368
|
Tocris Bioscience™
5182/10 |
10 mg |
£147.94
10mg |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18720509
|
Tocris Bioscience™
5182/50 |
50 mg |
£504.31
50mg |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Potent ROCK inhibitor (IC50 = 10 nM). Suppresses pMYPT1 and pCofilin levels in non-small cell lung carcinoma (NSCLC) cell lines. Also inhibits anchorage-independent growth of NSCLC cell lines in vitro.Specifications
>98% | |
OXA 06 dihydrochloride | |
OXA 06 dihydrochloride |
1825455-91-1 | |
404.31 |
For Research Use Only