Learn More
WTW™ OxiTop™-i Respirometric BOD Measuring System
The WTW™ OxiTop™-i Respirometric BOD Measuring System is designed for self-check BOD measurement with undiluted samples (according to DIN EN 1899 -2) at measuring ranges up to 4000 mg/L.
Brand: WTW™ 208272
322.05 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
- OxiTop™ is a respirometric vessel-top measuring system for BOD self-check measurement with undiluted samples (DIN EN 1899 -2)
- Measuring ranges up to 4000 mg/L BOD possible
- No need to convert data - direct input of sample volume
- Direct display of the BOD value on the measuring head screen
- Menu-operated, with backlit graphic LCD, control LED and operating buttons for direct BOD readings, volume pre-selection, measurement duration between 1 and 7 days selectable, display of measuring curve, display of current readings
Specifications
OxiTop™-i B | |
Backlit Graphic LCD | |
Measuring ranges up to 4000 mg/L BOD possible | |
One OxiTop™-i measuring head, blue |