Learn More
WTW™ OxiTop™-i Respirometric BOD Measuring System
The WTW™ OxiTop™-i Respirometric BOD Measuring System is designed for self-check BOD measurement with undiluted samples (according to DIN EN 1899 -2) at measuring ranges up to 4000 mg/L.
£339.00 - £6190.00
Specifications
Display Type | Backlit Graphic LCD |
---|---|
Range | Measuring ranges up to 4000 mg/L BOD possible |
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
16223561
|
WTW™
208276 |
OxiTop™-i 6 |
£1775.00
Each |
Estimated Shipment: 09-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16347231
|
WTW™
208272 |
OxiTop™-i B |
£339.00
Each |
Estimated Shipment: 09-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16273951
|
WTW™
208262 |
OxiTop™-i IS 12 |
£4645.00
Each |
Estimated Shipment: 14-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16219020
|
WTW™
208260 |
OxiTop™-i IS 6 |
£2695.00
Each |
Estimated Shipment: 28-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16308605
|
WTW™
208274 |
OxiTop™-i 2 |
£640.00
Each |
Estimated Shipment: 21-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16458763
|
WTW™
209700 |
OxiTop™-IDS 1 |
£453.00
Each |
Estimated Shipment: 09-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16391736
|
WTW™
209716 |
OxiTop™-IDS 12 |
£6190.00
Pack of 12 |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16367616
|
WTW™
208264 |
OxiTop™-i SET 2 |
£660.00
1 set |
Estimated Shipment: 23-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- OxiTop™ is a respirometric vessel-top measuring system for BOD self-check measurement with undiluted samples (DIN EN 1899 -2)
- Measuring ranges up to 4000 mg/L BOD possible
- No need to convert data - direct input of sample volume
- Direct display of the BOD value on the measuring head screen
- Menu-operated, with backlit graphic LCD, control LED and operating buttons for direct BOD readings, volume pre-selection, measurement duration between 1 and 7 days selectable, display of measuring curve, display of current readings
Specifications
Backlit Graphic LCD | |
Measuring ranges up to 4000 mg/L BOD possible |