Learn More
Thermo Scientific™ Oxoid™ Vitox Supplement
Oxoid Vitox Supplement is used for the preparation of Thayer Martin Medium and New York City Medium for the isolation of Neisseria spp.
£225.00 - £741.00
Specifications
Description | Vitox Supplement |
---|---|
Form | Freeze-dried |
Product Type | Microbiology Media |
Packaging Type | 2 x 5/Pk. |
Quantity | 5 x Freeze dried vial + 5 x Rehydration fluid |
Product Code | Brand | Yield | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Yield | Price | Quantity & Availability | |||||
12731870
|
Thermo Scientific™
SR0090A |
Each vial for 500mL medium |
£225.00
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13265709
|
Thermo Scientific™
SR0090H |
Each vial for 2L medium |
£741.00
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Oxoid Vitox Supplement is used for the preparation of Thayer Martin Medium and New York City Medium for the isolation of Neisseria spp.
- Contents: Vitox Supplement (5 vials) plus reconstitution fluid (5 vials)
- Add to GC Agar Base, Part No. CM0367B
Ordering Information:
- Each vial of Vitox plus reconstitution fluid supplements 500mL of medium
Includes:
- Five vials of Vitox plus five vials of reconstitution fluid
Specifications
Vitox Supplement | |
Microbiology Media | |
5 x Freeze dried vial + 5 x Rehydration fluid |
Freeze-dried | |
2 x 5/Pk. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.