Learn More
Chemical Identifiers
CAS | 118-75-2 |
---|---|
Molecular Formula | C6Cl4O2 |
Molecular Weight (g/mol) | 245.86 |
MDL Number | MFCD00001594 |
InChI Key | UGNWTBMOAKPKBL-UHFFFAOYSA-N |
Synonym | chloranil, p-chloranil, tetrachloro-p-benzoquinone, tetrachloro-1,4-benzoquinone, tetrachlorobenzoquinone, spergon, coversan, reranil, vulklor, 2,3,5,6-tetrachloro-1,4-benzoquinone |
PubChem CID | 8371 |
ChEBI | CHEBI:36703 |
IUPAC Name | 2,3,5,6-tetrachlorocyclohexa-2,5-diene-1,4-dione |
SMILES | ClC1=C(Cl)C(=O)C(Cl)=C(Cl)C1=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11425593
|
Thermo Scientific Alfa Aesar
A13495.14 |
25 g |
£28.04
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11435593
|
Thermo Scientific Alfa Aesar
A13495.22 |
100 g |
£60.40
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11445593
|
Thermo Scientific Alfa Aesar
A13495.36 |
500 g |
£224.12
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
p-Chloranil is used as a dye intermediate, oxidizing agent, vulcanizing agent and dehydrogenation reagent. It is also used to make chloranil electrodes for pH measurements. Further, it serves as a hydrogen acceptor and used for the aromatization reactions such as conversion of cyclohexadienes to the benzene derivatives. It is used to test the secondary amine. In addition to this, it is employed as a precursor to prepare diaziquone, which is used as a cancer chemotherapeutic agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsp-Chloranil is used as a dye intermediate, oxidizing agent, vulcanizing agent and dehydrogenation reagent. It is also used to make chloranil electrodes for pH measurements. Further, it serves as a hydrogen acceptor and used for the aromatization reactions such as conversion of cyclohexadienes to the benzene derivatives. It is used to test the secondary amine. In addition to this, it is employed as a precursor to prepare diaziquone, which is used as a cancer chemotherapeutic agent.
Solubility
Soluble in ether and acetone. Slightly soluble in chloroform, dimethylformamide, solvent naphtha, benzene, carbon disulfide and carbon tetrachloride. Insoluble in water, cold petroleum ether, methanol, dibutyl phthalate and cold alcohol.
Notes
Incompatible with strong oxidizing agents and strong bases.
Chemical Identifiers
118-75-2 | |
245.86 | |
UGNWTBMOAKPKBL-UHFFFAOYSA-N | |
8371 | |
2,3,5,6-tetrachlorocyclohexa-2,5-diene-1,4-dione |
C6Cl4O2 | |
MFCD00001594 | |
chloranil, p-chloranil, tetrachloro-p-benzoquinone, tetrachloro-1,4-benzoquinone, tetrachlorobenzoquinone, spergon, coversan, reranil, vulklor, 2,3,5,6-tetrachloro-1,4-benzoquinone | |
CHEBI:36703 | |
ClC1=C(Cl)C(=O)C(Cl)=C(Cl)C1=O |
Specifications
118-75-2 | |
1.97 | |
MFCD00001594 | |
393006 | |
chloranil, p-chloranil, tetrachloro-p-benzoquinone, tetrachloro-1,4-benzoquinone, tetrachlorobenzoquinone, spergon, coversan, reranil, vulklor, 2,3,5,6-tetrachloro-1,4-benzoquinone | |
UGNWTBMOAKPKBL-UHFFFAOYSA-N | |
2,3,5,6-tetrachlorocyclohexa-2,5-diene-1,4-dione | |
8371 | |
245.88 | |
p-Chloranil |
∼290°C (sublimation) | |
C6Cl4O2 | |
UN3077 | |
14,2078 | |
Soluble in ether and acetone. Slightly soluble in chloroform,dimethylformamide,solvent naphtha,benzene,carbon disulfide and carbon tetrachloride. Insoluble in water,cold petroleum ether,methanol,dibutyl phthalate and cold alcohol. | |
ClC1=C(Cl)C(=O)C(Cl)=C(Cl)C1=O | |
245.86 | |
CHEBI:36703 | |
97% |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P311-P332+P313-P362-P501c
H315-H319-H331
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'einecsNumber' : 204-274-4
missing translation for 'rtecsNumber' : DK6825000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- p-Chloranil
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P403-Store in a well-ventilated place.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : 2,3,5,6-Tetrachloro-2,5-cyclohexadiene-1,4-dione
RUO – Research Use Only