Learn More
p-Cymene, 99+%, Thermo Scientific Chemicals
£16.00 - £206.00
Chemical Identifiers
CAS | 99-87-6 |
---|---|
Molecular Formula | C10H14 |
Molecular Weight (g/mol) | 134.22 |
MDL Number | MFCD00008893 |
InChI Key | HFPZCAJZSCWRBC-UHFFFAOYSA-N |
Synonym | p-cymene, 4-isopropyltoluene, p-isopropyltoluene, dolcymene, p-cymol, paracymene, cymene, para-cymene, 1-isopropyl-4-methylbenzene, camphogen |
PubChem CID | 7463 |
ChEBI | CHEBI:28768 |
IUPAC Name | 1-methyl-4-propan-2-ylbenzene |
SMILES | CC1=CC=C(C=C1)C(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10447100
|
Thermo Scientific Acros
111762500 |
250 mL | Glass bottle |
£16.00
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10446910
|
Thermo Scientific Acros
111760010 |
1 L | Glass bottle |
£34.40
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10293940
|
Thermo Scientific Acros
111760025 |
2.5 L | Glass bottle |
£65.30
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10781535
|
Thermo Scientific Acros
111760100 |
10 L | Plastic drum |
£206.00
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
99-87-6 | |
134.22 | |
HFPZCAJZSCWRBC-UHFFFAOYSA-N | |
7463 | |
1-methyl-4-propan-2-ylbenzene |
C10H14 | |
MFCD00008893 | |
p-cymene, 4-isopropyltoluene, p-isopropyltoluene, dolcymene, p-cymol, paracymene, cymene, para-cymene, 1-isopropyl-4-methylbenzene, camphogen | |
CHEBI:28768 | |
CC1=CC=C(C=C1)C(C)C |
Specifications
99-87-6 | |
0.8540g/mL | |
47°C | |
99% min. (GC) | |
CH3C6H4CH(CH3)2 | |
MFCD00008893 | |
15, 2760 | |
p-cymene, 4-isopropyltoluene, p-isopropyltoluene, dolcymene, p-cymol, paracymene, cymene, para-cymene, 1-isopropyl-4-methylbenzene, camphogen | |
HFPZCAJZSCWRBC-UHFFFAOYSA-N | |
1-methyl-4-propan-2-ylbenzene | |
7463 | |
134.22 | |
Liquid |
-68.0°C | |
176.0°C to 178.0°C | |
Authentic | |
C10H14 | |
1.4890 to 1.4910 | |
05, II, 322 | |
0.854 | |
Solubility in water: practically insoluble. Other solubilities: soluble in alcohol,ether,chloroform,acetone,,benzene and ccl4 | |
CC1=CC=C(C=C1)C(C)C | |
134.22 | |
CHEBI:28768 | |
99+% | |
p-Cymene |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
If skin irritation occurs: Get medical advice/attentio
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-796-7
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 3
- Reproductive toxicity Category 2
Hazard Statement
- H226-Flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H331-Toxic if inhaled.
- H361-Suspected of damaging fertility or the unborn child.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P311-Call a POISON CENTER/doctor/ .
- P331-Do NOT induce vomiting.
RUO – Research Use Only