missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals p-Terphenyl, 99+%, pure Product Code.: 10513881

Thermo Scientific Chemicals p-Terphenyl, 99+%, pure

Product Code. 10513881
25 g, Plastic bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10513881

Brand: Thermo Scientific Chemicals 137920250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-94-4
Molecular Formula C18H14
Molecular Weight (g/mol) 230.31
MDL Number MFCD00003061
InChI Key XJKSTNDFUHDPQJ-UHFFFAOYSA-N
Synonym p-terphenyl, 1,1':4',1-terphenyl, p-diphenylbenzene, p-triphenyl, santowax p, 4-phenylbiphenyl, 4-phenyldiphenyl, para-terphenyl, biphenyl, 4-phenyl, pyrogallol tanninShow MoreShow Less
PubChem CID 7115
ChEBI CHEBI:52242
IUPAC Name 1,4-diphenylbenzene
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=CC=C3

Specifications

Melting Point 209.0°C to 216.0°C
Color White to Beige
Density 207g/mL
Boiling Point 383.0°C
Flash Point 207°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Linear Formula C6H5C6H4C6H5
Quantity 25 g
Beilstein 05, 695
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,benzene and toluene
Formula Weight 230.31
Percent Purity 99+%
Grade Pure
Physical Form Crystalline Powder, Shiny Crystals or Flakes
Chemical Name or Material p-Terphenyl, Pure
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467277 Certificate TypeCertificate of Analysis Date01/11/2024 Catalog Number137925000,137921000,137920250
Lot NumberA0466297 Certificate TypeCertificate of Analysis Date02/10/2024 Catalog Number137925000,137921000,137920250
Lot NumberA0464334 Certificate TypeCertificate of Analysis Date17/07/2024 Catalog Number137925000,137921000,137920250
Lot NumberA0461824 Certificate TypeCertificate of Analysis Date18/04/2024 Catalog Number137925000,137921000,137920250
Lot NumberA0458028 Certificate TypeCertificate of Analysis Date05/12/2023 Catalog Number137925000,137921000,137920250
compliance-icons
Product Identifier
  • p-Terphenyl
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : p-Terphenyl

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals p-Terphenyl, 99+%, pure >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.