missing translation for 'onlineSavingsMsg'
Learn More
p-Toluenesulfonic acid monohydrate, ACS reagent, Thermo Scientific Chemicals Product Code.: 15203356

p-Toluenesulfonic acid monohydrate, ACS reagent, Thermo Scientific Chemicals

Product Code. 15203356
100 g, Plastic bottle
Click to view available options
Quantity:
100 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 15203356

Brand: Thermo Scientific Acros 421211000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6192-52-5
Color White
Assay Percent Range 99%
Molecular Formula C7H10O4S
Linear Formula CH3C6H4SO3H·H2O
MDL Number MFCD00142137
Beilstein 11, 97
Fieser 01,1172; 04,508; 05,673; 06,597; 07,374; 08,488; 09,471; 11,535; 12,507
Merck Index 15, 9692
Synonym p-toluenesulfonic acid monohydrate, 4-methylbenzenesulfonic acid hydrate, 4-methylbenzenesulfonic acid monohydrate, p-toluene sulfonic acid monohydrate, unii-3bto78gaff, benzenesulfonic acid, 4-methyl-, monohydrate, ptoluenesulfonic acid, p-toluenesulfonic acid, monohydrate, 3bto78gaff, 4-methylbenzene-1-sulfonic acid hydrate
InChI Key KJIFKLIQANRMOU-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1)S(=O)(=O)O.O
IUPAC Name 4-methylbenzenesulfonic acid;hydrate
Molecular Weight (g/mol) 190.213
PubChem CID 521998
Formula Weight 190.22
Percent Purity ≥98.5% (Acid)
Grade ACS Reagent
Quantity 100 g
Packaging Plastic bottle
Physical Form Powder
Chemical Name or Material p-Toluenesulfonic acid monohydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465595 Certificate TypeCertificate of Analysis Date22/08/2024 Catalog Number421210250,421215000,421211000
Lot NumberA0464267 Certificate TypeCertificate of Analysis Date23/07/2024 Catalog Number421210250,421215000,421211000
Lot NumberA0457965 Certificate TypeCertificate of Analysis Date20/11/2023 Catalog Number421210250,421215000,421211000
Lot NumberA0440078 Certificate TypeCertificate of Analysis Date09/11/2023 Catalog Number421210250,421215000,421211000
Lot NumberA0455878 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number421210250,421215000,421211000
compliance-icons
Product Identifier
  • p-Toluenesulfonic acid monohydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
p-Toluenesulfonic acid monohydrate, ACS reagent, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.