Learn More
Sartorius Vivaspin™ 500 Centrifugal Concentrators
Provides a simple, one-step concentration procedure. Sartorius Vivaspin™ 500 Centrifugal Concentrators are disposable ultrafiltration devices for the concentration of biological samples with starting volumes up to 500μL. Available with PES membranes in a wide range of molecular weight cut-offs.
£119.00 - £860.00
Specifications
Disposable | Single-use |
---|---|
Material (Membrane) | PES |
Filtration Area | 64.5m m2 |
Housing Material | Polycarbonate |
Volume (Metric) | 500 μL |
Product Code | Brand | MWCO | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | MWCO | Quantity | Price | Quantity & Availability | ||||
10019460
|
Sartorius
VS0101 |
10 kDa | 25/Pk. |
£268.32
Pack of 25 |
In Stock
Log in to see stock. |
||||
10719001
|
Sartorius
VS0112 |
5 kDa | 100/Pk. |
£540.00
Pack of 100 |
Estimated Shipment: 08-10-2024
Log in to see stock. |
||||
10029510
|
Sartorius
VS0111 |
5 kDa | 25/Pk. |
£235.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
10125580
|
Sartorius
VS0102 |
10 kDa | 100/Pk. |
£814.00
Pack of 100 |
In Stock
Log in to see stock. |
||||
10681842
|
Sartorius
VS0121 |
30 kDa | 25/Pk. |
£244.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
10029470
|
Sartorius
VS0131 |
50 kDa | 25/Pk. |
£247.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
10375361
|
Sartorius
VS0141 |
100 kDa | 25/Pk. |
£250.78
Pack of 25 |
In Stock
Log in to see stock. |
||||
10459512
|
Sartorius
VS0132 |
50 kDa | 100/Pk. |
£580.00
Pack of 100 |
Estimated Shipment: 08-10-2024
Log in to see stock. |
||||
10073930
|
Sartorius
VS0122 |
30 kDa | 100/Pk. |
£860.00
Pack of 100 |
Estimated Shipment: 09-10-2024
Log in to see stock. |
||||
10499701
|
Sartorius
VS0151 |
300 kDa | 25/Pk. |
£238.00
Pack of 25 |
Estimated Shipment: 18-10-2024
Log in to see stock. |
||||
10223621
|
Sartorius
VS0142 |
100 kDa | 100/Pk. |
£799.00
Pack of 100 |
In Stock
Log in to see stock. |
||||
10767052
|
Sartorius
VS0152 |
300 kDa | 100/Pk. |
£575.00
Pack of 100 |
In Stock
Log in to see stock. |
||||
10727632
|
Sartorius
VS0161 |
1000 kDa | 25/Pk. | ||||||
10174281
|
Sartorius
VS0162 |
1000 kDa | 100/Pk. |
£343.72
Pack of 100 |
In Stock
Log in to see stock. |
||||
10184281
|
Sartorius
VS0171 |
0.2 μm | 25/Pk. |
£165.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
10658961
|
Sartorius
VS0172 |
0.2 μm | 100/Pk. |
£540.00
Pack of 100 |
In Stock
Log in to see stock. |
||||
10440282
|
Sartorius
VS0191 |
3 kDa | 25/Pk. |
£177.00
Pack of 25 |
In Stock
Log in to see stock. |
||||
10488742
|
Sartorius
VS01S1 |
5 each of 5K, 10K, 30K, 50K, 100K Da | 25/Pk. | ||||||
10689151
|
Sartorius
VS0192 |
3 kDa | 100/Pk. |
£575.00
Pack of 100 |
In Stock
Log in to see stock. |
||||
Description
Provides a simple, one-step concentration procedure. Sartorius Vivaspin™ 500 Centrifugal Concentrators are disposable ultrafiltration devices for the concentration of biological samples with starting volumes up to 500μL. Available with PES membranes in a wide range of molecular weight cut-offs.
- Patented vertical membrane design and thin channel filtration chamber minimizes membrane fouling and provides high speed concentrations
- Concentrate recoveries of > 90% can be reached in less than 30 minutes
- The dead stop pocket prevents concentration to dryness for safe sample concentration
- Universal rotor compatibility: can be used in fixed angle rotors accepting 2.2mL centrifuge tubes
- High-flux PES membranes for ultra-fast concentration
- Convenient concentrate recovery by pipetting
- Clear housing with volume graduation for easy reading/monitoring
Specifications
Single-use | |
64.5m m2 | |
500 μL |
PES | |
Polycarbonate | |
50 mm |