Learn More
Thermo Scientific™ Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with PES, CN, SFCA or Nylon Membranes
Achieve fast flow rates and high throughput for increased filtration efficiency with Nalgene Rapid-Flow sterile disposable bottle top filters.
£80.71 - £370.00
Specifications
Disposable | Single-use |
---|---|
Filter Type | Membrane Filter |
Material | Polystyrene |
Sterility | Sterile |
No. per Case | 12 |
Alert:
CAUTION: Use only sterile bottles designed for use with vacuum. Always use safety shield during vacuum procedures.
Product Code | Brand | Material (Membrane) | Volume (Metric) | Pore Size | Fits Bottle Neck Size (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material (Membrane) | Volume (Metric) | Pore Size | Fits Bottle Neck Size (Metric) | Price | Quantity & Availability | ||
10598282
|
Thermo Scientific™
596-4520 |
PES | 150 mL | 0.2 μm | 45mm |
£128.00
Pack of 12 |
In Stock
Log in to see stock. |
||
10743281
|
Thermo Scientific™
296-3345 |
PES | 150 mL | 0.45 μm | 33mm |
£111.00
Pack of 12 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
10319590
|
Thermo Scientific™
296-4545 |
PES | 150 mL | 0.45 μm | 45mm |
£181.00
Pack of 12 |
In Stock
Log in to see stock. |
||
10526952
|
Thermo Scientific™
595-3320 |
PES | 500 mL | 0.2 μm | 33mm |
£231.17
Pack of 12 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
10300461
|
Thermo Scientific™
595-4520 |
PES | 500 mL | 0.2 μm | 45mm |
£152.00
Pack of 12 |
In Stock
Log in to see stock. |
||
10706971
|
Thermo Scientific™
295-3345 |
PES | 500 mL | 0.45 μm | 33mm |
£226.00
Pack of 12 |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
10646421
|
Thermo Scientific™
295-4545 |
PES | 500 mL | 0.45 μm | 45mm |
£162.00
Pack of 12 |
In Stock
Log in to see stock. |
||
10491093
|
Thermo Scientific™
597-3320 |
PES | 1000 mL | 0.2 μm | 33mm |
£203.94
Pack of 12 |
Estimated Shipment: 05-11-2024
Log in to see stock. |
||
10401103
|
Thermo Scientific™
597-4520 |
PES | 1000 mL | 0.2 μm | 45mm |
£295.00
Pack of 12 |
In Stock
Log in to see stock. |
||
10352911
|
Thermo Scientific™
290-3320 |
SFCA | 150 mL | 0.2 μm | 33mm |
£80.71
Pack of 12 |
Estimated Shipment: 30-10-2024
Log in to see stock. |
||
10139560
|
Thermo Scientific™
290-4520 |
SFCA | 150 mL | 0.2 μm | 45mm |
£160.00
Pack of 12 |
In Stock
Log in to see stock. |
||
10027780
|
Thermo Scientific™
290-3345 |
SFCA | 150 mL | 0.45 μm | 33mm |
£112.00
Pack of 12 |
Estimated Shipment: 05-11-2024
Log in to see stock. |
||
10624281
|
Thermo Scientific™
290-4545 |
SFCA | 150 mL | 0.45 μm | 45mm |
£82.85
Pack of 12 |
In Stock
Log in to see stock. |
||
10037790
|
Thermo Scientific™
291-3320 |
SFCA | 500 mL | 0.2 μm | 33mm |
£118.00
Pack of 12 |
In Stock
Log in to see stock. |
||
10201371
|
Thermo Scientific™
291-4520 |
SFCA | 500 mL | 0.2 μm | 45mm |
£221.00
Pack of 12 |
In Stock
Log in to see stock. |
||
10529051
|
Thermo Scientific™
291-3345 |
SFCA | 500 mL | 0.45 μm | 33mm |
£112.72
Pack of 12 |
Estimated Shipment: 30-10-2024
Log in to see stock. |
||
10549241
|
Thermo Scientific™
291-4545 |
SFCA | 500 mL | 0.45 μm | 45mm |
£229.00
Pack of 12 |
In Stock
Log in to see stock. |
||
10139800
|
Thermo Scientific™
292-3320 |
SFCA | 1000 mL | 0.2 μm | 33mm |
£184.14
Pack of 12 |
Estimated Shipment: 30-10-2024
Log in to see stock. |
||
10790592
|
Thermo Scientific™
292-4520 |
SFCA | 1000 mL | 0.2 μm | 45mm |
£370.00
Pack of 12 |
In Stock
Log in to see stock. |
||
10441411
|
Thermo Scientific™
596-3320 |
PES | 150 mL | 0.2 μm | 33mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
Increase filtration efficiency. Thermo Scientific™ Nalgene™ Rapid-Flow™ single use bottle tops feature the exclusive Rapid-Flow support column design, providing fast flow rates and high throughput. The last line of defense against cell culture contamination—Nalgene Rapid-Flow bottle tops are the best choice for the filtration of cell culture media, serum, additives and buffers.
- Available in PES and SFCA membranes. Devices are color coded according to membrane.
- Devices feature the fastest flow rate and lowest clogging of comparable alternatives
- 0.1um PES available for mycoplasma protection
- 0.2um PES devices are stem cell and mouse embryonic assay (MEA) tested
- Non-pyrogenic and endotoxin free - Rapid-Flow filters meet the most rigorous testing standards for any vacuum filtration unit
- Available with Fit for Purpose Sterile documentation
Specifications
Single-use | |
Polystyrene | |
12 | |
Bottle Top Filter |
Membrane Filter | |
Sterile | |
12 | |
Sterile |