missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyrTrp Cell Dissociation Reagent
Recombinant and animal-derived component-free alternative to traditional porcine trypsin
£42.80 - £87.45
Specifications
Content And Storage | -20 °C |
---|---|
Product Type | Cell Culture Dissociation Reagent |
For Use With | Attachment-dependent cell types MRC-5, Vero, CHO K-1 cell lines grown with or without serum |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17205814
|
Cytiva
SV30209.01 |
100 mL |
£42.80
100mL |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17215814
|
Cytiva
SV30209.02 |
500 mL |
£87.45
500mL |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Animal-derived component-free (ADCF)
- Dye-free
- Suitable for a variety of applications and cell types
- Effective cell dissociation
- Does not impact cell line viability and doubling times
- For cell-based vaccine production and stem cell/primary cell/gene therapy cell culture processes
- For use with attachment-dependent cell types MRC-5, Vero, CHO K-1 cell lines grown with or without serum
Specifications
-20 °C | |
Cell Culture Dissociation Reagent | |
Attachment-dependent cell types MRC-5, Vero, CHO K-1 cell lines grown with or without serum |