missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pachymic acid, MedChemExpress
MedChemExpress Pachymic acid is a lanostrane-type triterpenoid from P. cocos. Pachymic acid inhibits Akt and ERK signaling pathways.
Brand: Medchem Express HY-N0371
This item is not returnable.
View return policy
Description
Pachymic acid is a lanostrane-type triterpenoid from P. cocos. Pachymic acid inhibits Akt and ERK signaling pathways.Specifications
Pachymic acid | |
Off-White | |
C33H52O5 | |
3-O-Acetyltumulosic acid | |
O[C@@H]([C@]1([H])[C@H](C(O)=O)CCC(C(C)C)=C)C[C@@]([C@]1(C)CC2)(C)C3=C2[C@]4(C)[C@](CC3)([H])C(C)(C)[C@@H](OC(C)=O)CC4 | |
528.76 | |
Research | |
Cancer-Kinase/protease |
29070-92-6 | |
Research | |
5 mg | |
DMSO : 10 mg/mL (18.91 mM; ultrasonic and warming and heat to 60°C) ∣Ethanol : 4.17 mg/mL (7.89 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble) | |
528.76 | |
98.0% | |
Solid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Research purposes only
Spot an opportunity for improvement?Share a Content Correction