Learn More
Pachymic acid, MedChemExpress
MedChemExpress Pachymic acid is a lanostrane-type triterpenoid from P. cocos. Pachymic acid inhibits Akt and ERK signaling pathways.
£246.84 - £1208.79
Specifications
Chemical Name or Material | Pachymic acid |
---|---|
CAS | 29070-92-6 |
Color | Off-White |
Purity Grade Notes | Research |
Molecular Formula | C33H52O5 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16458958
|
Medchem Express
HY-N0371 |
10 mg |
£384.78
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16451388
|
Medchem Express
HY-N0371 |
5 mg |
£246.84
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16404827
|
Medchem Express
HY-N0371 |
50 mg |
£1208.79
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pachymic acid is a lanostrane-type triterpenoid from P. cocos. Pachymic acid inhibits Akt and ERK signaling pathways.Specifications
Pachymic acid | |
Off-White | |
C33H52O5 | |
DMSO : 10 mg/mL (18.91 mM; ultrasonic and warming and heat to 60°C) ∣Ethanol : 4.17 mg/mL (7.89 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble) | |
528.76 | |
98.0% | |
Solid |
29070-92-6 | |
Research | |
3-O-Acetyltumulosic acid | |
O[C@@H]([C@]1([H])[C@H](C(O)=O)CCC(C(C)C)=C)C[C@@]([C@]1(C)CC2)(C)C3=C2[C@]4(C)[C@](CC3)([H])C(C)(C)[C@@H](OC(C)=O)CC4 | |
528.76 | |
Research | |
Cancer-Kinase/protease |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.