missing translation for 'onlineSavingsMsg'
Learn More
Palladium on activated carbon, 10% Pd, (50% wet with water for safety), unreduced, Thermo Scientific Chemicals Product Code.: 10697391

Palladium on activated carbon, 10% Pd, (50% wet with water for safety), unreduced, Thermo Scientific Chemicals

Product Code. 10697391
10 g, Glass bottle
Click to view available options
Quantity:
1 g
10 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10697391

Brand: Thermo Scientific Acros 422980100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-05-3, 7732-18-5, 7440-44-0
Molecular Formula Pd
Molecular Weight (g/mol) 106.42
MDL Number MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
InChI Key KDLHZDBZIXYQEI-UHFFFAOYSA-N
Synonym black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbonShow MoreShow Less
PubChem CID 23938
ChEBI CHEBI:33363
IUPAC Name palladium
SMILES [Pd]

Specifications

Color Black
Linear Formula Pd
Quantity 10 g
Fieser 01,778; 04,365; 05,498; 06,443; 07,274; 15,13; 16,356
Formula Weight 106.42
Percent Purity 10% (Pd)
Packaging Glass bottle
Form Powder
Chemical Name or Material Palladium on activated carbon

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0469226 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number422980250,422980010,422981000,422980100
Lot NumberA0459062 Certificate TypeCertificate of Analysis Date04/01/2024 Catalog Number422980250,422980010,422981000,422980100
Lot NumberA0455981 Certificate TypeCertificate of Analysis Date30/08/2023 Catalog Number422980250,422980010,422981000,422980100
Lot NumberA0452423 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number422980250,422980010,422981000,422980100
Lot NumberA0451761 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number422980250,422980010,422981000,422980100
Product Identifier
  • Palladium on activated carbon (50% wet with water for safety)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Palladium on activated carbon, 10% Pd, (50% wet with water for safety), unreduced, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.