missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Pallet Knives with Plastic Handles

These Pallet Knives have a hygienic stainless steel blade with an ergonomic black PVC handle.

Brand:  Sampling Systems™ A200-100

Additional Details : Weight : 0.00720kg

 View more versions of this product

Product Code. 15839591

  • £13.65 / Each
Estimated Shipment: 17-07-2024
to see stock.
Add to basket

Description

Description

  • Stainless steel blade with plastic PVC handle
  • Satin finish
  • Range of sizes available
Specifications

Specifications

Pallet Knife
Black Handle
190 mm
20 mm
Stainless Steel, Plastic
Satin
100 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers