missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Pam3CSK4, TLR1 and TLR2 Ligand
Applications: Functional, In vitro assay, In vivo assay, Ligand Activation
£145.00 - £184.00
Specifications
Concentration | 0.5 mg/mL |
---|---|
Content And Storage | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Use With (Application) | Activation of TLR1 and TLR2 heterodimer. Stimulation of TLR1 and TLR2 has been reported with 10 to 100ng/mL. Use in vitro assay reported in scientific literature (PMID: 24105263) Use in functional reported in scientific literature (PMID 25511699). Use in In-vivo reported in scientific literature (PMID 25466255). Use in ligand activation reported in multiple pieces scientific literature. |
Target | Pam3CSK4, TLR1 and TLR2 Ligand |
Species | Human/Mouse |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18705143
|
Novus Biologicals™
NBP2-25297-0.025mg |
0.025 mg |
£145.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18796653
|
Novus Biologicals™
NBP2-25297 |
0.1 mg |
£184.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pam3CSK4, TLR1 and TLR2 Ligand is a synthetic tripalmitoylated lipohexapeptide analog of the immunologically active N-terminal portion of bacterial lipoprotein. It activates monocytes and macrophages and is also a potent activator of proinflammatory transcription factor NF-kB. Pam3CSK4, TLR1 and TLR2 Ligand is recognized by a heterodimer formed between TLR1 and TLR2. Pam3CSK4, TLR1 and TLR2 Ligand stimulation: If your cell line does not naturally express TLR1+ TLR2, co-transfect with plasmids pCMV/TLR1and pCMV/TLR2. Forty-eight hours after transfection, stimulate cells with 10 to 100ng/ml of Pam3CSK4 for 6 to 24 hours. Determine Pam3CSK4 stimulation using appropriate detection assays. Buffer: Sterile waterSpecifications
0.5 mg/mL | |
Activation of TLR1 and TLR2 heterodimer. Stimulation of TLR1 and TLR2 has been reported with 10 to 100ng/mL. Use in vitro assay reported in scientific literature (PMID: 24105263) Use in functional reported in scientific literature (PMID 25511699). Use in In-vivo reported in scientific literature (PMID 25466255). Use in ligand activation reported in multiple pieces scientific literature. | |
Human/Mouse |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Pam3CSK4, TLR1 and TLR2 Ligand |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Pam3CSK4, TLR1 and TLR2 Ligand