missing translation for 'onlineSavingsMsg'
Learn More
Pamoic acid, 99% Product Code.: 11482218

Pamoic acid, 99%

Product Code. 11482218
1000 g
Click to view available options
Quantity:
1000 g
This item is not returnable. View return policy

Product Code. 11482218

Brand: Thermo Scientific Alfa Aesar A15207.0B

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 130-85-8 | C23H16O6 | 388.38 g/mol

Pamoic acid is used in preparation of repository derivatives of medicinal agents, used in pharmacology to increase the solubility of a drug in water.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pamoic acid is used in preparation of repository derivatives of medicinal agents, used in pharmacology to increase the solubility of a drug in water.

Solubility
Soluble in nitrobenzene, pyridine

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Specifications

CAS 130-85-8
Melting Point >300°C
Assay Percent Range 99%
Molecular Formula C23H16O6
MDL Number MFCD00004079
Beilstein 901319
Merck Index 14,7005
Synonym pamoic acid, embonic acid, 4,4 '-methylenebis 3-hydroxy-2-naphthoic acid, pamosaeure, unii-7 rrq8qz38n
Solubility Information Soluble in nitrobenzene, pyridine
InChI Key WLJNZVDCPSBLRP-UHFFFAOYSA-N
SMILES OC(=O)C1=C(O)C(CC2=C3C=CC=CC3=CC(C(O)=O)=C2O)=C2C=CC=CC2=C1
IUPAC Name 4-[(3-carboxy-2-hydroxynaphthalen-1-yl)methyl]-3-hydroxynaphthalene-2-carboxylic acid
Molecular Weight (g/mol) 388.38
PubChem CID 8546
ChEBI CHEBI:50186
Formula Weight 388.38
Percent Purity 99%
Quantity 1000 g
Chemical Name or Material Pamoic Acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462880 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA15207.30,A15207.18,A15207.0B
Lot NumberA0440874 Certificate TypeCertificate of Analysis Date22/04/2024 Catalog NumberA15207.30,A15207.18,A15207.0B
Lot NumberGBFA030284 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15207.30,A15207.18,A15207.0B
Lot NumberF5811A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15207.30,A15207.18,A15207.0B
Lot NumberFA030284 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15207.30,A15207.18,A15207.0B

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pamoic acid, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.