Learn More
Novus Biologicals™ PAP Pen Fine Tip
Brand:  Novus Biologicals™ NBP2-31111
203.20 GBP valid until 2025-03-28
Use promo code "25306" to get your promotional price.
Product Code. 18710404
  • £254.00 / Each
Explore more special offers
Specifications
Room Temperature
1 Each
Preparation of Working Solutions 1. Ready to be Used Directly on Wet Slides. Protocol Recommendations 1. Hold The Pen Point up and Press it with Your Fingertip to Take Blocked Air Out From The Pen Barrel to Prevent Overflow. 2. To Start, Press The Tip of The Pen Straight Down Until Fluid Starts Soaking The Tip and Then Draw a Circle Around The Specimen.
Tools and Accessories
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Novus Biologicals™ PAP Pen Fine Tip >