missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Papain, for Biochemistry
£40.60 - £802.00
Specifications
Name | Papain |
---|---|
pH Range | 4.8 to 6.2 |
Source | Plant |
Packaging | Glass bottle |
Form | Powder |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10657221
|
Thermo Scientific Chemicals
416765000 |
500g |
£802.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10701262
|
Thermo Scientific Chemicals
416760100 |
10g |
£40.60
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10400832
|
Thermo Scientific Chemicals
416761000 |
100g |
£202.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Papain | |
Plant | |
Powder |
4.8 to 6.2 | |
Glass bottle |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Papain
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
RUO – Research Use Only