missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Papain, Carica Papaya Latex Product Code.: 15414369
In Stock
to see stock.
Add to basket

Thermo Scientific Chemicals Papain, Carica Papaya Latex

Product Code. 15414369
100 mg
Click to view available options
Quantity:
25 mg
100 mg
1 g
This item is not returnable. View return policy

Product Code. 15414369

Brand: Thermo Scientific Chemicals J61875.MC

In Stock
to see stock.
Add to basket
Add to basket
Request Bulk or Custom Formats
This item is not returnable. View return policy

A cysteine protease; breaks down intercellular matrix of cartilage

Papain, from papaya latex, has been used to digest carotid artery smooth muscle cells. Papain is used in dissecting solutions. It is used to produce Fab fragments of antibodies. It is used for cell dissociation since it has been shown to be more effective and less damaging with certain tissues.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9001-73-4
Synonym EC 3.4.22.2

Specifications

For Use With (Application) A cysteine protease; breaks down intercellular matrix of cartilage
Molecular Weight (g/mol) 23 kd
Content And Storage 2°C to 8°C
Form Lyophilized powder,23 kDa
Quantity 100 mg
Product Type Protease

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS28K029 Certificate TypeCertificate of Analysis Date19/03/2025 Catalog NumberJ61875.03,J61875.MB,J61875.MC
Lot NumberP05L035 Certificate TypeCertificate of Analysis Date10/03/2025 Catalog NumberJ61875.03,J61875.MB,J61875.MC
Lot NumberN07L048 Certificate TypeCertificate of Analysis Date10/02/2025 Catalog NumberJ61875.03,J61875.MB,J61875.MC
Lot NumberM09L018 Certificate TypeCertificate of Analysis Date13/01/2025 Catalog NumberJ61875.03,J61875.MB,J61875.MC
Lot NumberX30K013 Certificate TypeCertificate of Analysis Date07/11/2024 Catalog NumberJ61875.03,J61875.MB,J61875.MC
compliance-icons compliance-icons
Product Identifier
  • Papain, Carica Papaya Latex
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Papain, Carica Papaya Latex

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Papain, Carica Papaya Latex >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.