Learn More
3M™ Particulate Filters
3M™ Particulate Filters can be used with 3M™ Gas and Vapour Filters 6000 Series to add particulate protection (in conjunction with a 501 filter retainer), and are designed to optimise your field of vision. Our filters are lightweight and feature a bayonet-style connection.
Brand:  3M™ 7000037673
29.35 GBP valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.12600kg
 View more versions of this product
Product Code. 15709017
  • £54.15 / 15 pairs
Estimated Shipment: 20-08-2024
to see stock.
Add to basket
Description
Description
  • Combines with the 6000 Series Gas Vapor cartridges to provide particulate and gas/vapor protection
  • Note: 3M 501 filter retainers needed to combine the 5000 and 6000 series
Specifications
Specifications
EN143 : 2000, CE Approved
P1R
Particulate Filter
6000 and 7000 Series face masks
Filter retainer
Electret
Solid and liquid particles
Filter