Learn More
DWK Life Sciences Pasteur pipettes made from Borosilikat glass 5.1
Pasteur pipettes are intended for single use in microbiological and serological laboratories
Brand: DWK Life Sciences 233302303
71.70 GBP valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 0.00300kg
Description
Thanks to their material properties, Pasteur pipettes made of borosilicate glass 5.1 are a high-quality alternative to plastic pipettes and pipettes made of soda-lime glass.
- Borosilicate glass 5.1 is more stable than soda-lime glass and is therefore insensitive to rapid temperature changes and chemical stress.
- Material properties ensure chemical resistance to acid and alkaline solutions
- Nearly inert borosilicate glass minimizes interactions with the material and reduces the risk of sample contamination
- Pasteur pipettes with short or long extended, fine tip (150 and 230 mm)
- The filling volume is approx. 2 ml.
- Natural rubber teat can be ordered separately.
Specifications
No | |
Borosilicate Glass (5.1) | |
4 x 250 |
6.95 mm | |
Single use in microbiological and serological laboratory applications; Pipetting liquids; Dropwise dosing of liquids and reagents; Distribution of heated liquid quantities | |
230 mm |