missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Pasteur pipettes made from Borosilikat glass 5.1
Pasteur pipettes are intended for single use in microbiological and serological laboratories
£79.65 - £96.30
Specifications
Autoclavable | No |
---|---|
Diameter (Metric) Outer | 6.95 mm |
Material | Borosilicate Glass (5.1) |
For Use With (Application) | Single use in microbiological and serological laboratory applications; Pipetting liquids; Dropwise dosing of liquids and reagents; Distribution of heated liquid quantities |
Quantity | 4 x 250 |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
15826881
|
DWK Life Sciences
233301504 |
150 mm |
£79.65
Pack of 1000 |
Estimated Shipment: 26-09-2024
Log in to see stock. |
|||||
15836881
|
DWK Life Sciences
233302303 |
230 mm |
£96.30
Pack of 1000 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
Description
Thanks to their material properties, Pasteur pipettes made of borosilicate glass 5.1 are a high-quality alternative to plastic pipettes and pipettes made of soda-lime glass.
- Borosilicate glass 5.1 is more stable than soda-lime glass and is therefore insensitive to rapid temperature changes and chemical stress.
- Material properties ensure chemical resistance to acid and alkaline solutions
- Nearly inert borosilicate glass minimizes interactions with the material and reduces the risk of sample contamination
- Pasteur pipettes with short or long extended, fine tip (150 and 230 mm)
- The filling volume is approx. 2 ml.
- Natural rubber teat can be ordered separately.
Specifications
No | |
Borosilicate Glass (5.1) | |
4 x 250 |
6.95 mm | |
Single use in microbiological and serological laboratory applications; Pipetting liquids; Dropwise dosing of liquids and reagents; Distribution of heated liquid quantities |