missing translation for 'onlineSavingsMsg'
Learn More
Patent Blue V, sodium salt, Thermo Scientific Chemicals Product Code.: 10439992

Patent Blue V, sodium salt, Thermo Scientific Chemicals

Product Code. 10439992
25 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10439992

Brand: Thermo Scientific Acros 339330250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20262-76-4
Molecular Formula C27H31N2NaO7S2
Molecular Weight (g/mol) 582.662
MDL Number MFCD09608138
InChI Key PMLFOMWMYRKZRF-UHFFFAOYSA-M
Synonym m-Hydroxytetraethyldiaminotriphenylcarbinol anhydride disulfonic acid, sodium salt, Acid Blue 3
PubChem CID 9916325
IUPAC Name sodium;4-[[4-(diethylamino)phenyl]-(4-diethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]-6-hydroxybenzene-1,3-disulfonateShow MoreShow Less
SMILES CCN(CC)C1=CC=C(C=C1)C(=C2C=CC(=[N+](CC)CC)C=C2)C3=CC(=C(C=C3S(=O)(=O)[O-])S(=O)(=O)[O-])O.[Na+]

Specifications

Dye Content 80% min.
Solubility Information Solubility in water: soluble.
Infrared Spectrum Authentic
Formula Weight 582.67
Packaging Glass bottle
Color Blue/Green
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Patent blue V, sodium salt

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465150 Certificate TypeCertificate of Analysis Date08/08/2024 Catalog Number339331000,339330050,339330250
Lot NumberA0463081 Certificate TypeCertificate of Analysis Date22/05/2024 Catalog Number339331000,339330050,339330250
Lot NumberA0457717 Certificate TypeCertificate of Analysis Date09/11/2023 Catalog Number339331000,339330050,339330250
Lot NumberA0455990 Certificate TypeCertificate of Analysis Date30/08/2023 Catalog Number339331000,339330050,339330250
Lot NumberA0392498 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number339331000,339330050,339330250
Product Identifier
  • Patent Blue V, sodium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Patent Blue V, sodium salt, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.