Learn More
Novacyt - Microgen Bioproducts PathFlow Rotavirus/Adenovirus Combi Test Cassette
The PathFlow Rota/Adeno Combi Test Cassette is a rapid chromatographic immunoassay for the qualitative detection of rotavirus and adenovirus in human faeces; the test utilises antibodies specific for rotavirus and adenovirus to selectively detect rotavirus and adenovirus from specimens.
Brand: Novacyt - Microgen Bioproducts M596CE
Description
- Diagnostic techniques such as electron microscopy (EM) and nucleic acid hybridisation are expensive and labour-intensive.
- The PathFlow™ Rota/Adeno Combi Test Cassette is a rapid chromatographic immunoassay for the qualitative detection of rotavirus and adenovirus in human faeces, providing results from 10 minutes.
Specifications
ISO13485 ISO9001 | |
Yes | |
Genogroup 1 and Genogroup 2 Antibodies | |
Swab | |
Test Strip | |
Specimen Collection/ Stool Container, Timer, Centrifuge and Pipette | |
Plastic | |
Animal | |
25 Tests and 1 Bottle (1 mL) | |
Norovirus | |
Eukaryota |
2°C to 30°C | |
Non-waived | |
25 Tests | |
0.957 | |
A Qualitative, Lateral Flow Immunoassay for The Detection of Norovirus in Human Faeces Specimens | |
25 Tests | |
1/Test | |
Carton with sealed foil pouches | |
Contains Genogroup 1 and Genogroup 2 monoclonal antibody coated particles and Genogroup1 and Genogroup 2 monoclonal antibodies coated on the membrane | |
0.917 | |
15 minutes |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.