missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ eXpress c-Ret-GFRα2 Functional Assay
Click to view available options
Quantity:
1,000 dp (10 x 96-well)
Unit Size:
1 set
Description
The PathHunter™ eXpress c-Ret-GFRα2 Functional Assay provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting RTKs. This kit assesses RTK activation via detection of activated c-Ret and GFRα2 dimerization-induced SH2 domain protein recruitment. This product uses EFC technology. The included cells overexpress PK-tagged c-Ret and an EA-tagged SH2 domain protein. It also overexpresses an untagged coreceptor, GFRα2 Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Cyno version of the protein.
Specifications
Specifications
| Assay Entry | Kinase |
| For Use With (Application) | Assay-Ready eXpress Kits |
| Quantity | 1,000 dp (10 x 96-well) |
| Target Entry | c-Ret-GFRα2 |
| Target | c-Ret-GFRα2 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction