missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ eXpress FGR Activity Assay
Click to view available options
Quantity:
1,000 dp (10 x 96-well)
Unit Size:
1 set
Description
The PathHunter™ eXpress FGR Activity Assay provides an easy-to-use cell based assay to study small molecule drug candidates targeting CTKs. This kit assesses CTK activation via detection of resulting GHR phosphorylation-induced SH2 domain protein recruitment, mediated by FGR-JAK2 fusion protein. This product uses EFC technology. The included cells overexpress PK-tagged relevant cytokine receptor and EA-tagged SH2 domain protein alongside an engineered fusion of the cytokine receptor binding domain of JAK2 (Janus Kinase 2) to the catalytic domain from the CTK of interest, FGR. Upon activation of the cytokine receptor, the CRBD-CTK fusion phosphorylates the receptor, leading to SH2-EA recruitment and production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Specifications
Specifications
| Assay Entry | SH2 recruitment |
| For Use With (Application) | Assay-Ready eXpress Kits |
| Primary Agonist | Recombinant Human Growth Hormone |
| Quantity | 1,000 dp (10 x 96-well) |
| Target Entry | FGR |
| Cell Line | U2OS |
| Target | FGR |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction