missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Intelsius™ PathoSeal™ 95
The PathoSeal™ 95 is a flexible secondary packaging solution certified to withstand a pressure difference of 95 kPa in order to comply with UN3373 Biological Substance, Category B regulations.
Brand: Intelsius™ BZ015
This item is not returnable.
View return policy
Description
- Suitable for use within Category B packaging solutions
- Tamper evident seal
- Simple close system for fast sample collection
- Uniquely numbered for sample traceability
- External document pouch for relevant paperwork
- Certified to withstand 95 kPa
Specifications
300 mL Absorbent | |
A3 | |
394 x 310 mm (15.5 x 12.2 in.) |
UN3373 | |
Sample Transport Bag |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction