missing translation for 'onlineSavingsMsg'
Learn More

Intelsius™ PathoSeal™ 95

The PathoSeal™ 95 is a flexible secondary packaging solution certified to withstand a pressure difference of 95 kPa in order to comply with UN3373 Biological Substance, Category B regulations.

£82.49 - £507.26

Specifications

Certifications/Compliance UN3373
Type Sample Transport Bag
View More Specs

Products 4
Product Code Brand Size Dimensions (L x W) Price Quantity & Availability  
Product Code Brand Size Dimensions (L x W) Price Quantity & Availability  
17404612
Encompass_Preferred
View Documents
Intelsius™
BZ013
A6 139 x 99 mm (5.5 x 3.9 in.)
£389.81
Pack of 500
View alternative products
Save Up to 
»
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
 
15604886
Encompass_Preferred
View Documents
Intelsius™
BZ008
A5 201 x 152 mm (7.9 x 6.0 in.)
£82.49
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment: 08-01-2025
to see stock.
Add to basket
 
17494602
Encompass_Preferred
View Documents
Intelsius™
BZ009
A4 284 x 200 mm (11.2 x 7.9 in.)
£152.96
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
 
17414612
Encompass_Preferred
View Documents
Intelsius™
BZ015
A3 394 x 310 mm (15.5 x 12.2 in.)
£507.26
Pack of 250
View alternative products
Save Up to 
»
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
 
Description

Description

  • Suitable for use within Category B packaging solutions
  • Tamper evident seal
  • Simple close system for fast sample collection
  • Uniquely numbered for sample traceability
  • External document pouch for relevant paperwork
  • Certified to withstand 95 kPa
Specifications

Specifications

UN3373
Sample Transport Bag
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.