Learn More
Intelsius™ PathoSorb Absorbent Pouch
Protect and separate sample vials with this PathoSorb Absorbent sample Pouch.
Brand: Intelsius™ AB005
Additional Details : Weight : 8.00000kg
Description
- Keeps sample tubes safe and secure
- Lint free
- Suitable in supporting UN3373 Category B and UN2900/UN2814 Category A transport regulations
Specifications
4 Bays | |
Absorbent Sample Pouch | |
152 mL |
UN3373, UN2900, UN2814 | |
225 x 125 mm (8.9 x 4.9 in.) |