Learn More
Intelsius™ PathoSorb Absorbent Pouch
Protect and separate sample vials with this PathoSorb Absorbent sample Pouch.
Brand:  Intelsius™ AB008
Encompass_Preferred
 View more versions of this product
Product Code. 17328589
  • £129.40 / Pack of 200
Estimated Shipment: 02-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Keeps sample tubes safe and secure
  • Lint free
  • Suitable in supporting UN3373 Category B and UN2900/UN2814 Category A transport regulations
Specifications
Specifications
6 Bays
Absorbent Sample Pouch
215 mL
UN3373, UN2900, UN2814
325 x 125 mm (12.8 x 4.9 in.)