missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences CD81 Isolation Kit for FABian™
IBA Lifesciences™ CD81 Isolation Kit for FABian™ is for reversible cell isolation delivering label-free target cells.
£84.14 - £664.86
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Donor Source | Human Whole Blood |
Sufficient For | 9 mL Whole Blood |
Description | Isolation of label-free cells |
Product Type | Cell Isolation Kit |
Description
- The kit is used for the selection of PBMCs from whole blood or buffy coat in conjunction with the FABian™ system.
- It includes CD81 Fab-Strep (human), the Cell Isolation Column, Cell Isolation Buffer, Biotin for Cell Elution and tubes and a luer lock adapter with the syringe.
- Each kit is sufficient for one FABian™ run to isolate cells from 9mL whole blood.
Specifications
2°C to 8°C | |
9 mL Whole Blood | |
Cell Isolation Kit |
Human Whole Blood | |
Isolation of label-free cells |