missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Phosphate Buffered Saline (PBS) Tablets
Phosphate Buffered Saline Tablets (PBS) are a balanced salt mixture for easy PBS buffer preparation for a variety of applications, including reagent preparation, diluting cells for flow cytometry, and cell culture reagent.
Brand: MP Biomedicals™ 092810306
166.55 GBP valid until 2024-12-31
Use promo code "21951" to get your promotional price.
This item is not returnable.
View return policy
Description
- No need for weighing and mixing chemicals
- Simply dissolve PBS tablets in water
- One tablet dissolved in 100 mL water yields buffer containing 0.01M phosphate, 0.0027M KCl, and 0.137M NaCl, pH 7.4 at 25°C
- Research use only
Specifications
No Calcium, No Magnesium | |
200 Tablets | |
MFCD00011360 | |
[Cl-].[K+] | |
74.55 |
Phosphate Buffered Saline (PBS) | |
ClK | |
WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
potassium chloride | |
Tablet |