Learn More
Thermo Scientific™ PCC-54™ Detergent Concentrate
A laboratory cleaning solution for labware utensils, surface materials and glassware, including beakers, pipets, lenses, mirrors; RBS-35 alternative.
Brand: Thermo Scientific™ 72288
Code : PN
Additional Details : CAS Number : 7320-34-5 Weight : 3.00000kg Transport : UN number : 3267 Chem class : 8 Pack group : III
Description
Thermo Scientific™ PCC-54 Detergent Concentrate is a general cleaning agent for use on many laboratory surface materials and glassware, including lenses, mirrors, and reflectors.
Highlights:
- Removal of blood residue from glass or plastic beakers and flasks Colored dyes from porcelain mortar and pestles HIV virus from glassware, tabletops and flooring For removing radioisotopes on contaminated tabletops
- Rinses clean, providing film-free surfaces Safe to handle, and PCC-Pfree does not contain phosphates that can harm the environment Use a 2%- 20% solution at 50°C for soaking stubborn biological residues away
Specifications
Liquid | |
PCC-PFree, 2%- 20% Solution | |
PCC-54 Detergent Concentrate | |
3 L |
PCC-54™ | |
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20 to 25°C. | |
1 Ea. |
Safety and Handling
- PCC-54 Detergent Concentrate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : tetrapotassium pyrophosphate, sodium carbonate, Isotridecanol, ethoxylated, Sodium hypochlorite, 4-N
For Research Use Only. Not for use in diagnostic procedures.