Learn More
PCK1 Mouse anti-Mouse, Unconjugated, Clone: 2F11B5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66862-1-IG-20UL
This item is not returnable.
View return policy
Description
The PCK1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PCK1. This antibody recognizes mouse, rat, pig, human antigen. The PCK1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
PCK1 | |
Monoclonal | |
760 μg/mL | |
Mouse | |
Primary | |
Mouse, Rat, Pig, Human |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
2F11B5 | |
Unconjugated | |
20 μL | |
5105 | |
IgG1 |