missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PCK1 Mouse anti-Mouse, Unconjugated, Clone: 2F11B5, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
Antigen | PCK1 |
---|---|
Clone | 2F11B5 |
Concentration | 760 μg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16829125
|
Proteintech
66862-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16819125
|
Proteintech
66862-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The PCK1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PCK1. This antibody recognizes mouse, rat, pig, human antigen. The PCK1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
PCK1 | |
760 μg/mL | |
Monoclonal | |
Mouse | |
5105 | |
Primary |
2F11B5 | |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
Unconjugated | |
Mouse, Rat, Pig, Human | |
IgG1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title