missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ PCR Product Pre-Sequencing Kit
The PCR Product Pre-Sequencing Kit uses a novel, enzymatic clean-up method to pre-treat PCR products prior to sequencing, without any subsequent purification or separation steps.
£79.25 - £1108.00
Specifications
Content And Storage | Store at -5 to -30°C. |
---|---|
For Use With (Application) | PCR Purification |
Product Line | ExoSAP-IT™ |
Protein Form | Recombinant |
Research Category | Genetic Diseases |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
15637748
|
Applied Biosystems™
70995.1.KT |
100 |
£79.25
100 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15647748
|
Applied Biosystems™
70996.1.KT |
500 Reactions |
£319.00
500 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15657748
|
Applied Biosystems™
70997.1.KT |
2000 |
£1108.00
2000 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Remove unincorporated primers and nucleotides in about 5 minutes
- Prepare DNA sequencing samples with one pipetting step
- Conserve precious PCR products with up to 100% recovery, regardless of amplicon length
- Minimize spin column or bead clean-up
Specifications
Store at -5 to -30°C. | |
ExoSAP-IT™ | |
Genetic Diseases |
PCR Purification | |
Recombinant | |
Dry Ice |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Applied Biosystems™ PCR Product Pre-Sequencing Kit