Learn More
Tocris Bioscience™ PD 98059
MEK inhibitor
Brand:  Tocris Bioscience™ 1213/1
73.48 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18073852
  • £94.00 / 1mg
Explore more special offers
Description
Description
PD 98059 is an inhibitor of mitogen-activated protein kinase kinase (MKK/ MEK). Acts by binding to the inactivated form of MEK, thereby preventing its phosphorylation by cRAF or MEK kinase (IC50 = 2-7 μM). Inhibits cell growth and proliferation in acute myelogenous leukemia (AML) cell lines; causes G1 arrest by blocking p53-dependent p21 induction. Enhances embryonic stem cell self-renewal.
Specifications
Specifications
98%
167869-21-8
1 mg
M.W. (Observed): 267.28
2-(2-Amino-3-methoxyphenyl)-4 H-1-benzopyran-4-one
Room Temperature
PD 98059
PD 98059