Learn More
Cytiva PD MidiTrap™ G-25 Sample Preparation Columns
Use for cleanup of biomolecules with a molecular weight >5000 Mr in sample volumes up to 1mL. Cytiva Life Sciences™ PD MidiTrap™ G-25 Sample Preparation Columns are convenient and prepacked.
Brand: Cytiva 28-9180-08
Code : GE
Additional Details : Weight : 0.75060kg
Includes: Four centrifuge adapters
Description
- Developed for a wide range of applications: desalting, buffer exchange, removal of low-molecular weight compounds, etc.
- Equally suited for gravity flow or centrifugation protocols
- Accommodates sample volumes of 0.5 to 1.0mL for gravity, 0.75 to 1.0mL with centrifugation
- High recoveries of 70 to 90%, depending on biomolecule
- High desalting capacityof >90%
Specifications
PD MidiTrap Sephadex G-25 gravity column | |
0.51 in. | |
Each | |
26mm | |
Four centrifuge adapters, instructions | |
2 to 13 | |
Cross-linked Dextran | |
Gravity Column |
8.5mL | |
13mm | |
1.02 in. | |
>100cm/hr. | |
Polyethylene | |
50μm in dry form, 38 to 235μm in wet form | |
4° to 30°C | |
Convenient and rapid cleanup with high reproducibility and the potential for high throughput by running multiple samples in parallel |
Safety and Handling
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : 5-chloro-2-methyl-4-isothiazolin- 3-one,2-methyl-2H-isothiazol-3-one