missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PDGFRA [T674I] Recombinant Human Protein

PDGFRA T674I is a Gleevec resistant form of PDGFRA.

Brand:  Invitrogen™ PV3847

Code : 50

Additional Details : Weight : 0.05250kg

 View more versions of this product

Product Code. 11563577

  • £434.00 / 10µg
Estimated Shipment: 05-08-2024
to see stock.

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

PDGFRA is a cell surface receptor tyrosine kinase which binds members of the platelet-derived growth factor family.

Specifications

Specifications

NP_006197, T674T
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
89.8kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Recombinant, Cytoplasmic
platelet-derived growth factor receptor, alpha polypeptide
aa 550-1089; T674I
Recombinant
Baculovirus
Receptor Tyrosine Kinases
Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation
5156
10μg
RUO - research use only
PDGFRA T674I, PDGFR alpha
PDGFR Kinase Family
Human
GST Tag
Kinases & Inhibitors
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers