missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PDGFRA [T674I] Recombinant Human Protein
PDGFRA T674I is a Gleevec resistant form of PDGFRA.
£434.00 - £12060.00
Specifications
Accession Number | NP_006197, T674T |
---|---|
Formulation | 50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol |
Molecular Weight (g/mol) | 89.8kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Regulatory Status | RUO - research use only |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11563577
|
Invitrogen™
PV3847 |
10μg |
£434.00
10µg |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15498573
|
Invitrogen™
PR7385B |
100μg |
£2555.00
100µg |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10494193
|
Invitrogen™
PV4118 |
1mg |
£12060.00
1mg |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
PDGFRA is a cell surface receptor tyrosine kinase which binds members of the platelet-derived growth factor family.
Specifications
NP_006197, T674T | |
89.8kDa | |
RUO - research use only | |
PDGFR Kinase Family | |
Recombinant | |
Baculovirus |
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
PDGFRA T674I, PDGFR alpha | |
Human | |
GST Tag |