missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PDGFRA [T674I] Recombinant Human Protein

PDGFRA T674I is a Gleevec resistant form of PDGFRA.

Brand:  Invitrogen™ PR7385B

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15498573

  • £2555.00 / 100µg
Estimated Shipment: 05-08-2024
to see stock.

Description

Description

PDGFRA is a cell surface receptor tyrosine kinase which binds members of the platelet-derived growth factor family.

Specifications

Specifications

NP_006197, T674T
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
100μg
RUO - research use only
PDGFRA T674I, PDGFR alpha
Human
GST Tag
Frozen
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
89.8kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Cytoplasmic
PDGFR Kinase Family
Recombinant
Baculovirus
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers