missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18262-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This antibody is specific to PDK1.
Serine/threonine kinase that plays a key role in regulation of glucose and fatty acid metabolism and homeostasis via phosphorylation of the pyruvate dehydrogenase subunits PDHA1 and PDHA2. This inhibits pyruvate dehydrogenase activity, and thereby regulates metabolite flux through the tricarboxylic acid cycle, down-regulates aerobic respiration and inhibits the formation of acetyl-coenzyme A from pyruvate. Plays an important role in cellular responses to hypoxia and is important for cell proliferation under hypoxia. Protects cells against apoptosis in response to hypoxia and oxidative stress.Specifications
PDK1 | |
Polyclonal | |
Unconjugated | |
PDK1 | |
PDH kinase 1, PDHK1, PDK1, Pyruvate dehydrogenase kinase isoform 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116551, 228026, 5163 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q15118, Q63065, Q8BFP9 | |
PDK1 | |
PDK1 Fusion Protein Ag13121 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |